Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011893.3 | NP_036023.3 | 1 - 2 | 117 | 70 | |
GenBank mRNA | AK089455.1 | - | 1 - 2 | 48 | 70 | |
AK155898.1 | - | 1 - 2 | 117 | 70 | ||
L14543.1 | - | 1 - 2 | 51 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons |