Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145858.1 | NP_001139330.1 | 7 - 8 | 812 | 103 | |
NM_001145859.1 | NP_001139331.1 | 7 - 8 | 815 | 103 | ||
NM_011893.3 | NP_036023.3 | 7 - 8 | 712 | 103 | ||
GenBank mRNA | AK089455.1 | - | 7 - 8 | 643 | 103 | |
AK143548.1 | - | 7 - 8 | 817 | 103 | ||
AK150265.1 | - | 7 - 8 | 814 | 103 | ||
AK155103.1 | - | 7 - 8 | 669 | 103 | ||
AK155898.1 | - | 7 - 8 | 712 | 103 | ||
AK155970.1 | - | 7 - 8 | 760 | 103 | ||
BC010198.1 | - | 7 - 8 | 650 | 3967543 | 103 | |
BC085497.1 | - | 7 - 8 | 703 | 5696296 | 103 | |
L14543.1 | - | 7 - 8 | 646 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |