Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289803.1 | NP_001276732.1 | 11 - 12 | 1599 | 84 | |
NM_001289804.1 | NP_001276733.1 | 12 - 13 | 1648 | 84 | ||
NM_181857.4 | NP_862905.1 | 11 - 12 | 1599 | 84 | ||
GenBank mRNA | AK133063.1 | - | 12 - 13 | 1568 | 84 | |
AY135562.3 | - | 11 - 12 | 1521 | 84 | ||
BC141101.1 | - | 11 - 12 | 1499 | 9055713 | 84 | |
BC145378.1 | - | 11 - 12 | 1499 | 9053936 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |