Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chchd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190322.2 | NP_001177251.1 | 2 - 3 | 352 | 63 | |
NM_001190323.2 | NP_001177252.1 | 2 - 3 | 228 | 63 | ||
NM_001190324.2 | NP_001177253.1 | 2 - 3 | 183 | 63 | ||
NM_001285804.1 | NP_001272733.1 | 2 - 3 | 383 | 63 | ||
NM_181391.4 | NP_852056.1 | 2 - 3 | 397 | 63 | ||
NR_104354.1 | 2 - 3 | 397 | 63 | |||
NR_104355.1 | 2 - 3 | 397 | 63 | |||
NR_104356.1 | 1 - 2 | 108 | 63 | |||
GenBank mRNA | AA980189.1 | - | 2 - 3 | 180 | 63 | |
AI851035.1 | - | 3 - 4 | 63 | |||
AK003308.1 | - | 2 - 3 | 229 | 63 | ||
AK007844.1 | - | 2 - 3 | 372 | 63 | ||
AK013200.1 | - | 2 - 3 | 370 | 63 | ||
AK145105.1 | - | 2 - 3 | 372 | 63 | ||
AK168647.1 | - | 2 - 3 | 327 | 63 | ||
AK169018.1 | - | 2 - 3 | 372 | 63 | ||
BC030366.1 | - | 2 - 3 | 355 | 5370748 | 63 | |
BX527959.1 | - | 2 - 3 | 144 | 63 | ||
CB057699.1 | - | 2 - 3 | 309 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |