Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clvs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028940.2 | NP_083216.1 | 2 - 3 | 782 | 61 | |
GenBank mRNA | AK016622.1 | - | 2 - 3 | 706 | 61 | |
AK032549.1 | - | 2 - 3 | 771 | 61 | ||
AK084350.1 | - | 2 - 3 | 778 | 61 | ||
BC062923.1 | - | 2 - 3 | 737 | 6848546 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |