Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkain3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290410.1 | NP_001277339.1 | 4 - 5 | 825 | 86 | |
GenBank mRNA | AK053823.1 | - | 4 - 5 | 710 | 86 | |
AK141456.1 | - | 4 - 5 | 725 | 86 | ||
EF058052.1 | - | 4 - 5 | 472 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |