Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpy19l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081201.1 | NP_001074670.1 | 1 - 2 | 128 | 146 | |
GenBank mRNA | AK144417.1 | - | 1 - 2 | 80 | 146 | |
AK172656.1 | - | 1 - 2 | 136 | 146 | ||
BC147500.1 | - | 1 - 2 | 84 | 9056363 | 146 | |
BC157932.1 | - | 1 - 2 | 84 | 9088105 | 146 | |
BC157988.1 | - | 1 - 2 | 84 | 9088143 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |