Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK041962.1 | - | 9 - 10 | 1011 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |