Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081397.1 | NP_001074866.1 | 16 - 17 | 2212 | 114 | |
GenBank mRNA | AB429291.1 | - | 14 - 15 | 1820 | 114 | |
AK082350.1 | - | 16 - 17 | 1948 | 114 | ||
BC082311.1 | - | 16 - 17 | 2078 | 114 | ||
BC151049.1 | - | 16 - 17 | 2166 | 9087962 | 114 | |
BC151051.1 | - | 16 - 17 | 2167 | 9087964 | 114 | |
BC157966.1 | - | 16 - 17 | 2167 | 9007290 | 114 | |
BC172122.1 | - | 16 - 17 | 2167 | 9054408 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |