Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081397.1 | NP_001074866.1 | 1 - 2 | 322 | 105 | |
GenBank mRNA | AK082350.1 | - | 1 - 2 | 58 | 105 | |
BC072580.1 | - | 1 - 2 | 334 | 105 | ||
BC082311.1 | - | 1 - 2 | 190 | 105 | ||
BC151049.1 | - | 1 - 2 | 276 | 9087962 | 105 | |
BC151051.1 | - | 1 - 2 | 277 | 9087964 | 105 | |
BC157966.1 | - | 1 - 2 | 277 | 9007290 | 105 | |
BC172122.1 | - | 1 - 2 | 277 | 9054408 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |