Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153391.2 | NP_700440.2 | 11 - 12 | 1193 | 59 | |
GenBank mRNA | AK043836.1 | - | 11 - 12 | 1150 | 59 | |
AK052303.1 | - | 11 - 12 | 1173 | 59 | ||
AK129413.1 | - | 3 - 4 | 282 | 59 | ||
AK147970.1 | - | 11 - 12 | 1193 | 59 | ||
AK166172.1 | - | 11 - 12 | 1159 | 59 | ||
AY029258.1 | - | 11 - 12 | 1263 | 59 | ||
BC060134.1 | - | 11 - 12 | 1187 | 59 | ||
BC082301.1 | - | 10 - 11 | 1073 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |