Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbpj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080927.2 | NP_001074396.1 | 3 - 4 | 365 | 80 | |
NM_001080928.1 | NP_001074397.1 | 3 - 4 | 251 | 80 | ||
NM_001277116.1 | NP_001264045.1 | 5 - 6 | 447 | 80 | ||
NM_009035.5 | NP_033061.3 | 3 - 4 | 267 | 80 | ||
GenBank mRNA | AK028363.1 | - | 5 - 6 | 425 | 80 | |
AK080359.1 | - | 3 - 4 | 160 | 80 | ||
AK090006.1 | - | 3 - 4 | 153 | 80 | ||
AK132145.1 | - | 3 - 4 | 209 | 80 | ||
AK139087.1 | - | 3 - 4 | 152 | 80 | ||
AK145187.1 | - | 3 - 4 | 159 | 80 | ||
AK152746.1 | - | 3 - 4 | 268 | 80 | ||
AK153163.1 | - | 4 - 5 | 155 | 80 | ||
AK184128.1 | - | 2 - 3 | 129 | 80 | ||
BC035299.1 | - | 1 - 2 | 53 | 80 | ||
BC051387.1 | - | 3 - 4 | 196 | 4460290 | 80 | |
BY155397.1 | - | 3 - 4 | 394 | 80 | ||
BY716845.1 | - | 3 - 4 | 338 | 80 | ||
CD540070.1 | - | 3 - 4 | 205 | 80 | ||
S63463.1 | - | 3 - 4 | 268 | 80 | ||
X17459.1 | - | 3 - 4 | 263 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |