Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136058.2 | NP_001129530.1 | 7 - 8 | 1125 | 68 | |
NM_007765.4 | NP_031791.3 | 7 - 8 | 990 | 68 | ||
GenBank mRNA | AB006714.1 | - | 7 - 8 | 962 | 68 | |
AK158793.1 | - | 7 - 8 | 1069 | 68 | ||
AK159049.1 | - | 7 - 8 | 934 | 68 | ||
BC031738.1 | - | 7 - 8 | 919 | 4505284 | 68 | |
BC065046.1 | - | 7 - 8 | 1103 | 5686818 | 68 | |
U72875.1 | - | 7 - 8 | 735 | 68 | ||
Y09080.1 | - | 7 - 8 | 965 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |