Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam53a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178390.3 | NP_848477.2 | 1 - 2 | 295 | 78 | |
GenBank mRNA | AK010537.1 | - | 1 - 2 | 232 | 78 | |
AK086386.1 | - | 1 - 2 | 260 | 78 | ||
AK155077.1 | - | 1 - 2 | 249 | 78 | ||
AK163326.1 | - | 1 - 2 | 266 | 78 | ||
BB848576.1 | - | 1 - 2 | 296 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |