Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtcap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027221.3 | NP_081497.2 | 1 - 2 | 110 | 82 | |
GenBank mRNA | AK007999.1 | - | 1 - 2 | 82 | ||
BC027812.1 | - | 1 - 2 | 108 | 5251276 | 82 | |
BC107024.1 | - | 1 - 2 | 87 | 40049838 | 82 | |
BC107025.1 | - | 1 - 2 | 87 | 40049842 | 82 | |
CK129285.1 | - | 1 - 2 | 110 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |