Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003933.2 | NP_001003933.1 | 5 - 6 | 2900 | 87 | |
NM_001003934.2 | NP_001003934.1 | 6 - 7 | 2957 | 87 | ||
NM_001271486.1 | NP_001258415.1 | 6 - 7 | 1994 | 87 | ||
NM_001271487.1 | NP_001258416.1 | 5 - 6 | 833 | 87 | ||
NM_053076.3 | NP_444306.1 | 4 - 5 | 776 | 87 | ||
GenBank mRNA | AB046114.1 | - | 4 - 5 | 770 | 87 | |
AF195940.1 | - | 4 - 5 | 720 | 87 | ||
AK032109.1 | - | 6 - 7 | 1971 | 87 | ||
AK036892.1 | - | 4 - 5 | 742 | 87 | ||
AK049845.1 | - | 4 - 5 | 708 | 87 | ||
AK088670.1 | - | 4 - 5 | 708 | 87 | ||
AK146505.1 | - | 4 - 5 | 584 | 87 | ||
AK148792.1 | - | 4 - 5 | 735 | 87 | ||
AK148947.1 | - | 4 - 5 | 737 | 87 | ||
AK165683.1 | - | 4 - 5 | 707 | 87 | ||
AK194472.1 | - | 80 | 87 | |||
AK217412.1 | - | 3 - 4 | 333 | 87 | ||
AK218971.1 | - | 3 - 4 | 342 | 87 | ||
AY164700.1 | - | 4 - 5 | 770 | 87 | ||
AY427822.1 | - | 5 - 6 | 2881 | 87 | ||
AY507126.1 | - | 4 - 5 | 568 | 87 | ||
AY507127.1 | - | 4 - 5 | 2503 | 87 | ||
AY750849.1 | - | 6 - 7 | 2938 | 87 | ||
BC005478.1 | - | 4 - 5 | 679 | 87 | ||
BC014697.1 | - | 4 - 5 | 687 | 87 | ||
BC036717.1 | - | 4 - 5 | 685 | 87 | ||
BC079882.1 | - | 6 - 7 | 2867 | 6839323 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |