Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noc3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021315.2 | NP_067290.2 | 17 - 18 | 2093 | 82 | |
GenBank mRNA | AB077991.1 | - | 17 - 18 | 2056 | 82 | |
AF233884.1 | - | 6 - 7 | 556 | 82 | ||
AK134051.1 | - | 17 - 18 | 2052 | 82 | ||
AK168944.1 | - | 17 - 18 | 2096 | 82 | ||
BC031132.1 | - | 17 - 18 | 2056 | 4954951 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |