Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028102.1 | NP_082378.1 | 11 - 12 | 1437 | 77 | |
GenBank mRNA | AK079311.1 | - | 10 - 11 | 1418 | 77 | |
AK173010.1 | - | 9 - 10 | 1112 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |