Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009619.4 | NP_033749.2 | 9 - 10 | 909 | 87 | |
GenBank mRNA | BC060975.1 | - | 9 - 10 | 872 | 87 | |
BC129920.1 | - | 9 - 10 | 875 | 87 | ||
X64227.1 | - | 9 - 10 | 919 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |