Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009069.4 | NP_033095.1 | 1 - 2 | 109 | 79 | |
NR_028056.1 | 1 - 2 | 109 | 79 | |||
GenBank mRNA | AK014126.1 | - | 1 - 2 | 100 | 79 | |
AK018785.1 | - | 1 - 2 | 57 | 79 | ||
AK164196.1 | - | 1 - 2 | 100 | 79 | ||
AK168254.1 | - | 1 - 2 | 68 | 79 | ||
BC012694.1 | - | 1 - 2 | 69 | 4023732 | 79 | |
CV562852.1 | - | 1 - 2 | 48 | 79 | ||
U71205.1 | - | 1 - 2 | 46 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |