Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172709.3 | NP_766297.2 | 2 - 3 | 557 | 106 | |
GenBank mRNA | AF548337.1 | - | 2 - 3 | 557 | 106 | |
AK040792.1 | - | 2 - 3 | 544 | 106 | ||
AK041023.1 | - | 2 - 3 | 331 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |