Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027949.1 | NP_082225.1 | 3 - 4 | 677 | 141 | |
GenBank mRNA | AK005852.1 | - | 3 - 4 | 677 | 141 | |
AK148191.1 | - | 3 - 4 | 760 | 141 | ||
AK155352.1 | - | 3 - 4 | 478 | 141 | ||
BC049113.1 | - | 3 - 4 | 774 | 141 | ||
BC057174.1 | - | 3 - 4 | 524 | 5004450 | 141 | |
BC132517.1 | - | 3 - 4 | 467 | 40130794 | 141 | |
BC138571.1 | - | 3 - 4 | 467 | 8861591 | 141 | |
BC145337.2 | - | 3 - 4 | 467 | 9053893 | 141 | |
BC145338.1 | - | 3 - 4 | 467 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |