Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027949.1 | NP_082225.1 | 4 - 5 | 774 | 118 | |
GenBank mRNA | AK005852.1 | - | 4 - 5 | 774 | 118 | |
AK155352.1 | - | 4 - 5 | 575 | 118 | ||
BC049113.1 | - | 4 - 5 | 871 | 118 | ||
BC057174.1 | - | 4 - 5 | 621 | 5004450 | 118 | |
BC132517.1 | - | 4 - 5 | 564 | 40130794 | 118 | |
BC138571.1 | - | 4 - 5 | 564 | 8861591 | 118 | |
BC145337.2 | - | 4 - 5 | 564 | 9053893 | 118 | |
BC145338.1 | - | 4 - 5 | 564 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |