Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270996.1 | NP_001257925.1 | 13 - 14 | 1534 | 68 | |
NM_007404.2 | NP_031430.2 | 13 - 14 | 1534 | 68 | ||
GenBank mRNA | AK122188.1 | - | 13 - 14 | 1517 | 68 | |
AK148174.1 | - | 13 - 14 | 1420 | 68 | ||
AK155582.1 | - | 13 - 14 | 1495 | 68 | ||
AK155672.1 | - | 13 - 14 | 1427 | 68 | ||
AK161778.1 | - | 13 - 14 | 1457 | 68 | ||
BC047156.1 | - | 13 - 14 | 1412 | 6488707 | 68 | |
BC052710.1 | - | 13 - 14 | 1406 | 68 | ||
D50412.1 | - | 2 - 3 | 118 | 68 | ||
U06145.1 | - | 1 - 2 | 100 | 68 | ||
U41765.1 | - | 13 - 14 | 1406 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |