Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Whsc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK079952.1 | - | 2 - 3 | 298 | 67 | |
AK156746.1 | - | 5 - 6 | 1537 | 67 | ||
AK170040.1 | - | 5 - 6 | 1600 | 67 | ||
BC036336.1 | - | 4 - 5 | 1129 | 67 | ||
BC064447.1 | - | 5 - 6 | 1575 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |