Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153572.2 | NP_705800.1 | 1 - 2 | 124 | 83 | |
GenBank mRNA | AK049620.1 | - | 1 - 2 | 478 | 83 | |
BC030434.1 | - | 1 - 2 | 111 | 5369313 | 83 | |
BE653455.1 | - | 1 - 2 | 124 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |