Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163493.1 | NP_001156965.1 | 8 - 9 | 2443 | 84 | |
NM_146258.2 | NP_666370.3 | 8 - 9 | 2386 | 84 | ||
GenBank mRNA | AK050259.1 | - | 8 - 9 | 2444 | 84 | |
AK136202.1 | - | 4 - 5 | 969 | 84 | ||
BC027830.1 | - | 8 - 9 | 2398 | 5320966 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |