Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081001.2 | NP_001074470.1 | 15 - 16 | 7888 | 141 | |
NM_009765.3 | NP_033895.2 | 15 - 16 | 7694 | 141 | ||
GenBank mRNA | AK165522.1 | - | 5 - 6 | 1023 | 141 | |
BC059904.1 | - | 5 - 6 | 1151 | 141 | ||
U65594.1 | - | 15 - 16 | 7671 | 141 | ||
U82270.1 | - | 14 - 15 | 7380 | 141 | ||
U89652.1 | - | 14 - 15 | 7418 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |