Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293768.1 | NP_001280697.1 | 1 - 2 | 517 | 58 | |
GenBank mRNA | AK209495.1 | - | 1 - 2 | 46 | 58 | |
AY486147.1 | - | 1 - 2 | 470 | 58 | ||
BC025640.1 | - | 1 - 2 | 173 | 5322622 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |