Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400012224.mm.1 |
NM_032008.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012224.mm.1 |
TC1400001621.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012204.mm.1 |
NM_032008.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012204.mm.1 |
TC1400001621.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012208.mm.1 |
NM_032008.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012208.mm.1 |
TC1400001621.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012210.mm.1 |
NM_032008.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012210.mm.1 |
TC1400001621.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012219.mm.1 |
NM_032008.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012219.mm.1 |
TC1400001621.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012217.mm.1 |
NM_032008.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012217.mm.1 |
TC1400001621.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012221.mm.1 |
NM_032008.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012221.mm.1 |
TC1400001621.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400012207.mm.1 |
NM_032008.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400012207.mm.1 |
TC1400001621.mm.1 |
MTA TC |