Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bod1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081422.2 | NP_001074891.1 | 11 - 12 | 7968 | 92 | |
GenBank mRNA | AF013969.1 | - | 2 - 3 | 3427 | 92 | |
AK122495.1 | - | 2 - 3 | 3112 | 92 | ||
AK163093.1 | - | 2 - 3 | 1066 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |