Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapre3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133350.1 | NP_579928.1 | 2 - 3 | 271 | 82 | |
GenBank mRNA | AF468005.1 | - | 2 - 3 | 271 | 82 | |
AK177097.1 | - | 2 - 3 | 186 | 82 | ||
BC048833.1 | - | 2 - 3 | 273 | 82 | ||
BC057918.1 | - | 2 - 3 | 271 | 5134461 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |