Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tkt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009388.6 | NP_033414.1 | 6 - 7 | 888 | 82 | |
GenBank mRNA | AF195533.1 | - | 5 - 6 | 554 | 82 | |
AK002627.1 | - | 6 - 7 | 841 | 82 | ||
AK012794.1 | - | 6 - 7 | 843 | 82 | ||
AK030446.1 | - | 6 - 7 | 871 | 82 | ||
AK140965.1 | - | 6 - 7 | 872 | 82 | ||
AK144146.1 | - | 6 - 7 | 843 | 82 | ||
AK145677.1 | - | 6 - 7 | 841 | 82 | ||
AK146157.1 | - | 6 - 7 | 841 | 82 | ||
AK150139.1 | - | 6 - 7 | 841 | 82 | ||
AK150769.1 | - | 6 - 7 | 837 | 82 | ||
AK150844.1 | - | 6 - 7 | 837 | 82 | ||
AK150856.1 | - | 6 - 7 | 837 | 82 | ||
AK150901.1 | - | 6 - 7 | 841 | 82 | ||
AK151916.1 | - | 6 - 7 | 836 | 82 | ||
AK151969.1 | - | 6 - 7 | 836 | 82 | ||
AK152245.1 | - | 6 - 7 | 836 | 82 | ||
AK152460.1 | - | 6 - 7 | 857 | 82 | ||
AK152886.1 | - | 6 - 7 | 841 | 82 | ||
AK159922.1 | - | 6 - 7 | 841 | 82 | ||
AK166763.1 | - | 6 - 7 | 875 | 82 | ||
AK167084.1 | - | 6 - 7 | 875 | 82 | ||
AK191384.1 | - | 2 - 3 | 166 | 82 | ||
AK214732.1 | - | 2 - 3 | 165 | 82 | ||
BC055336.1 | - | 6 - 7 | 815 | 6413263 | 82 | |
U05809.1 | - | 6 - 7 | 836 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |