Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172526.3 | NP_766114.1 | 2 - 3 | 733 | 82 | |
GenBank mRNA | AK039884.1 | - | 2 - 3 | 734 | 82 | |
BC147554.1 | - | 1 - 2 | 437 | 9056545 | 82 | |
BC147555.1 | - | 1 - 2 | 437 | 9056546 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |