Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnisr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025669.1 | NP_079945.1 | 8 - 9 | 1677 | 111 | |
GenBank mRNA | AK012036.1 | - | 8 - 9 | 1219 | 111 | |
AK017507.1 | - | 8 - 9 | 1204 | 111 | ||
AK020035.1 | - | 8 - 9 | 1210 | 111 | ||
AK031964.1 | - | 8 - 9 | 1213 | 111 | ||
AK160288.1 | - | 8 - 9 | 1204 | 114 | ||
BC141031.1 | - | 8 - 9 | 1541 | 9055643 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |