Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146097.3 | NP_666209.1 | 6 - 7 | 614 | 116 | |
NR_033744.1 | 6 - 7 | 681 | 116 | |||
GenBank mRNA | AK076027.1 | - | 6 - 7 | 569 | 116 | |
AK132291.1 | - | 6 - 7 | 600 | 116 | ||
BC018472.1 | - | 6 - 7 | 556 | 3661842 | 116 | |
BC053745.1 | - | 6 - 7 | 649 | 6505520 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |