Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033573.2 | NP_291051.1 | 4 - 5 | 1402 | 68 | |
GenBank mRNA | AF273750.1 | - | 4 - 5 | 1423 | 68 | |
AK078394.1 | - | 4 - 5 | 1422 | 68 | ||
BC027831.1 | - | 4 - 5 | 1435 | 68 | ||
BC052072.1 | - | 4 - 5 | 1402 | 5717482 | 68 | |
BC082784.1 | - | 4 - 5 | 1420 | 4191155 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |