Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175013.2 | NP_778178.3 | 6 - 7 | 1091 | 113 | |
GenBank mRNA | AK035507.1 | - | 6 - 7 | 1091 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |