Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcrls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030707.3 | NP_109632.2 | 3 - 4 | 367 | 152 | |
GenBank mRNA | AB028174.1 | - | 3 - 4 | 398 | 152 | |
AF302046.1 | - | 3 - 4 | 436 | 152 | ||
AF329486.1 | - | 3 - 4 | 368 | 152 | ||
AK034125.1 | - | 3 - 4 | 368 | 152 | ||
AK076123.1 | - | 3 - 4 | 401 | 152 | ||
AY506556.1 | - | 3 - 4 | 407 | 152 | ||
AY506557.1 | - | 3 - 4 | 407 | 152 | ||
BC016551.1 | - | 3 - 4 | 410 | 3602803 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |