Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277149.1 | NP_001264078.1 | 29 - 30 | 6420 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |