Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277149.1 | NP_001264078.1 | 9 - 10 | 3222 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |