Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010698.4 | NP_034828.3 | 7 - 8 | 1148 | 103 | |
GenBank mRNA | AB221652.1 | - | 7 - 8 | 895 | 103 | |
BC079611.1 | - | 7 - 8 | 1041 | 30360903 | 103 | |
U89487.1 | - | 7 - 8 | 1037 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |