Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081093.2 | NP_001074562.1 | 2 - 3 | 310 | 71 | |
NM_001293801.1 | NP_001280730.1 | 2 - 3 | 310 | 71 | ||
GenBank mRNA | BC132291.1 | - | 2 - 3 | 219 | 40130568 | 71 |
BC144758.1 | - | 2 - 3 | 219 | 9053306 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |