Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100034960.mm.1 |
NM_011486.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034960.mm.1 |
NM_213659.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034960.mm.1 |
NM_213660.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034960.mm.1 |
TC1100003790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034965.mm.1 |
NM_011486.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034965.mm.1 |
NM_213659.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034965.mm.1 |
NM_213660.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034965.mm.1 |
TC1100003790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034975.mm.1 |
NM_011486.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034975.mm.1 |
NM_213659.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034975.mm.1 |
NM_213660.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034975.mm.1 |
TC1100003790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034953.mm.1 |
NM_011486.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034953.mm.1 |
NM_213659.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034953.mm.1 |
NM_213660.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034953.mm.1 |
TC1100003790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034957.mm.1 |
NM_011486.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034957.mm.1 |
NM_213659.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034957.mm.1 |
NM_213660.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034957.mm.1 |
TC1100003790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034961.mm.1 |
NM_011486.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034961.mm.1 |
NM_213659.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034961.mm.1 |
NM_213660.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034961.mm.1 |
TC1100003790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034951.mm.1 |
NM_011486.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034951.mm.1 |
NM_213659.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034951.mm.1 |
NM_213660.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034951.mm.1 |
TC1100003790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034972.mm.1 |
NM_011486.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034972.mm.1 |
NM_213659.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034972.mm.1 |
NM_213660.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034972.mm.1 |
TC1100003790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034949.mm.1 |
NM_011486.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034949.mm.1 |
NM_213659.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034949.mm.1 |
NM_213660.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034949.mm.1 |
TC1100003790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034974.mm.1 |
NM_011486.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034974.mm.1 |
NM_213659.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034974.mm.1 |
NM_213660.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034974.mm.1 |
TC1100003790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034950.mm.1 |
NM_011486.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034950.mm.1 |
NM_213659.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034950.mm.1 |
NM_213660.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034950.mm.1 |
TC1100003790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034971.mm.1 |
NM_011486.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034971.mm.1 |
NM_213659.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034971.mm.1 |
NM_213660.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034971.mm.1 |
TC1100003790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034969.mm.1 |
NM_011486.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034969.mm.1 |
NM_213659.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034969.mm.1 |
NM_213660.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034969.mm.1 |
TC1100003790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034952.mm.1 |
NM_213659.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034952.mm.1 |
NM_213660.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034952.mm.1 |
TC1100003790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034963.mm.1 |
NM_011486.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034963.mm.1 |
NM_213659.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034963.mm.1 |
NM_213660.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034963.mm.1 |
TC1100003790.mm.1 |
MTA TC |