Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400013067.mm.1 |
NM_019975.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013067.mm.1 |
NM_019975.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013067.mm.1 |
TC1400002859.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400013071.mm.1 |
NM_019975.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013071.mm.1 |
NM_019975.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013071.mm.1 |
TC1400002859.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400013068.mm.1 |
NM_019975.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013068.mm.1 |
NM_019975.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013068.mm.1 |
TC1400002859.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400013073.mm.1 |
NM_019975.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013073.mm.1 |
NM_019975.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013073.mm.1 |
TC1400002859.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400013064.mm.1 |
NM_019975.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013064.mm.1 |
NM_019975.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013064.mm.1 |
TC1400002859.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400013069.mm.1 |
NM_019975.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013069.mm.1 |
NM_019975.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400013069.mm.1 |
TC1400002859.mm.1 |
MTA TC |