Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pglyrp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165968.1 | NP_001159440.1 | 4 - 5 | 527 | 69 | |
NM_207263.2 | NP_997146.2 | 4 - 5 | 527 | 69 | ||
GenBank mRNA | AY518699.1 | - | 4 - 5 | 481 | 69 | |
BC120840.1 | - | 4 - 5 | 522 | 40129763 | 69 | |
BC144902.1 | - | 4 - 5 | 527 | 9053450 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |