Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2q1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027315.4 | NP_081591.3 | 1 - 2 | 428 | 70 | |
GenBank mRNA | AK146707.1 | - | 1 - 2 | 241 | 70 | |
AY945937.1 | - | 1 - 2 | 325 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |