Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010252.4 | NP_034382.2 | 2 - 3 | 654 | 114 | |
GenBank mRNA | AK032128.1 | - | 2 - 3 | 339 | 114 | |
AK034085.1 | - | 2 - 3 | 242 | 114 | ||
AK034332.1 | - | 2 - 3 | 655 | 114 | ||
AK038468.1 | - | 2 - 3 | 391 | 114 | ||
AK039068.1 | - | 2 - 3 | 352 | 114 | ||
AK047752.1 | - | 2 - 3 | 404 | 114 | ||
AK049956.1 | - | 2 - 3 | 392 | 114 | ||
AK140399.1 | - | 2 - 3 | 404 | 114 | ||
AK163541.1 | - | 2 - 3 | 362 | 114 | ||
BC099939.1 | - | 2 - 3 | 363 | 30090762 | 114 | |
X55272.1 | - | 2 - 3 | 437 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |