Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039195.1 | NP_001034284.1 | 6 - 7 | 1292 | 76 | |
NM_001083806.1 | NP_001077275.1 | 6 - 7 | 1292 | 76 | ||
NM_013540.2 | NP_038568.2 | 6 - 7 | 1292 | 76 | ||
GenBank mRNA | AB109210.1 | - | 6 - 7 | 885 | 76 | |
AB109211.1 | - | 6 - 7 | 885 | 76 | ||
AB111957.1 | - | 6 - 7 | 885 | 76 | ||
AK014389.1 | - | 6 - 7 | 1337 | 76 | ||
AK043490.1 | - | 6 - 7 | 1265 | 76 | ||
AK044574.1 | - | 6 - 7 | 1224 | 76 | ||
AK046861.1 | - | 6 - 7 | 1292 | 76 | ||
AK086614.1 | - | 6 - 7 | 1226 | 76 | ||
AK134572.1 | - | 6 - 7 | 1222 | 76 | ||
AK134647.1 | - | 5 - 6 | 738 | 76 | ||
AK134772.1 | - | 5 - 6 | 863 | 76 | ||
AK163941.1 | - | 6 - 7 | 1285 | 76 | ||
BC048248.1 | - | 6 - 7 | 1151 | 6494013 | 76 | |
X57498.1 | - | 6 - 7 | 1087 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |