Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162999.2 | NP_001156471.1 | 7 - 8 | 785 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |